1-Bromo-8-methoxynaphthalene 95%

Supplier: Ambeed
Warning

A610642-100MG A610642-1G A610642-250MG A610642-5G
77117-370EA 51.99 USD
77117-370 77117-372 77117-374 77117-376
1-Bromo-8-methoxynaphthalene 95%
1-Bromo-8-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
MW: 237.09 g/mol
MDL Number: MFCD18410561
CAS Number: 83710-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR