BAM 7 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-ethoxyphenyl)hydrazone], 4-(2-(2-Ethoxyphenyl)hydrazono)-5-methyl-2-(4-phenylthiazol-2-yl)-2,4-dihydro-3H-pyrazol-3-one, Bax Activator Molecule 7

A608237-10MG A608237-1MG A608237-25MG A608237-50MG
77117-350EA 82.4 USD
77117-350 77117-352 77117-354 77117-356
BAM 7 98%
BAM 7

Precaución: for Research Use Only

Formula: C₂₁H₁₉N₅O₂S
MW: 405.476 g/mol
MDL Number: MFCD01246556
CAS Number: 331244-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR