1-(p-Tolyl)ethylamine 98%

Supplier: Ambeed
Danger

Synonyms: 4-(1-Aminoethyl)toluene, 1-(4-Methylphenyl)ethylamine

A607741-25G A607741-5G
77117-338EA 128.37 USD
77117-338 77117-340
1-(p-Tolyl)ethylamine 98%
1-(p-Tolyl)ethylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD02177111
CAS Number: 586-70-9
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR