Ethyl-2,4,6-trimethylbenzoate 95%

Supplier: Ambeed
Warning

A605282-1G A605282-25G
77117-296EA 17.51 USD
77117-296 77117-298
Ethyl-2,4,6-trimethylbenzoate 95%
Ethyl-2,4,6-trimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MDL Number: MFCD00015439
CAS Number: 1754-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR