Gliclazide 98%

Supplier: Ambeed
Warning

Synonyms: N-((Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)carbamoyl)-4-methylbenzenesulphonamide, 1-(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)-3-[(4-methylphenyl)sulphonyl]urea, 1-[3-Azabicyclo[3.3.0]oct-3-yl]-3-p-toluenesulphonylurea

A604981-100G A604981-1G A604981-1MG A604981-25G A604981-5G
77117-278EA 79.57 USD
77117-278 77117-280 77117-282 77117-284 77117-286
Gliclazide 98%
Gliclazide

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₃O₃S
MW: 323.41 g/mol
MDL Number: MFCD00409893
CAS Number: 21187-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR