6-Bromo-4-hydroxy-2-methylquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-methylquinolin-4-ol

A602132-10G A602132-1G A602132-5G A602132-25G
77117-266EA 186.72 USD
77117-266 77117-268 77117-270 77686-526
6-Bromo-4-hydroxy-2-methylquinoline 98%
6-Bromo-4-hydroxy-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.081 g/mol
MDL Number: MFCD00484433
CAS Number: 103030-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR