2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

A599323-10G A599323-1G A599323-250MG
77117-250EA 57.68 USD
77117-250 77117-252 77117-254
2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride 95%
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃O₂S
MDL Number: MFCD00052912
CAS Number: 175205-54-6
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR