4-(4-Methoxyphenyl)piperidine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A594080-250MG
A594080-5G
A594080-1G
77117-236EA
55.62
USD
77117-236
77117-238
77686-506
4-(4-Methoxyphenyl)piperidine hydrochloride 95%
4-(4-Methoxyphenyl)piperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₂H₁₈ClNO |
MDL Number:
MFCD02178921 CAS Number: 6748-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...