N-(4-((6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-yl)oxy)benzo[d]thiazol-2-yl)acetamide 98+%

Supplier: Ambeed
Warning

A593913-10MG A593913-1MG A593913-5MG
77117-230EA 42.22 USD
77117-230 77117-232 77117-234
N-(4-((6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-yl)oxy)benzo[d]thiazol-2-yl)acetamide 98+%
N-(4-((6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-yl)oxy)benzo[d]thiazol-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₂₀H₁₃F₃N₄O₂S
MDL Number: MFCD14584859
CAS Number: 659730-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR