1,1,2-Trimethylhydrazine dihydrochloride 97%

Supplier: Ambeed
Danger

A584593-1G A584593-25G
77117-198EA 26.78 USD
77117-198 77117-200
1,1,2-Trimethylhydrazine dihydrochloride 97%
1,1,2-Trimethylhydrazine dihydrochloride

Precaución: for Research Use Only

Formula: C₃H₁₂Cl₂N₂
MW: 147.04 g/mol
MDL Number: MFCD07369993
CAS Number: 339539-94-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR