Wogonin 98%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dihydroxy-8-methoxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxy-8-methoxyflavone

A583686-100MG A583686-10MG A583686-1MG A583686-50MG A583686-5MG A583686-1G
77117-186EA 125.19 USD
77117-186 77117-188 77117-190 77117-192 77117-194 77686-483
Wogonin 98%
Wogonin

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₅
MW: 284.3 g/mol
MDL Number: MFCD00017736
CAS Number: 632-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR