Ethyl-2-bromoisovalerate 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromoisovaleric Acid Ethyl Ester, 2-Bromo-3-methylbutyric Acid Ethyl Ester, Ethyl 2-bromo-3-methylbutanoate, Ethyl 2-Bromo-3-methylbutyrate

A581854-25G A581854-5G
77117-174EA 14.86 USD
77117-174 77117-176
Ethyl-2-bromoisovalerate 98%
Ethyl-2-bromoisovalerate

Precaución: for Research Use Only

Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
MDL Number: MFCD00026855
CAS Number: 609-12-1
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR