3-[4-(Trifluoromethyl)phenyl]propan-1-ol 96%

Supplier: Ambeed
Warning

Synonyms: 3-(4-(Trifluoromethyl)phenyl)propan-1-ol, 4-(3-Hydroxyprop-1-yl)benzotrifluoride

A581583-1G A581583-250MG
77117-170EA 132.62 USD
77117-170 77117-172
3-[4-(Trifluoromethyl)phenyl]propan-1-ol 96%
3-[4-(Trifluoromethyl)phenyl]propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃O
MW: 204.189 g/mol
MDL Number: MFCD09028744
CAS Number: 180635-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR