tert-Butyl ((3R,4S)-3-hydroxypiperidin-4-yl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A574020-100MG
A574020-250MG
77117-148EA
134.74
USD
77117-148
77117-150
tert-Butyl ((3R,4S)-3-hydroxypiperidin-4-yl)carbamate 98%
tert-Butyl ((3R,4S)-3-hydroxypiperidin-4-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₃ MW: 216.27 g/mol |
MDL Number:
MFCD18834105 CAS Number: 642478-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...