2,4,6-Triiodophenol 98%

Supplier: Ambeed
Warning

A573565-1G A573565-5G A573565-100G A573565-25G
77117-142EA 8.49 USD
77117-142 77117-144 77686-461 77686-462
2,4,6-Triiodophenol 98%
2,4,6-Triiodophenol

Precaución: for Research Use Only

Formula: C₆H₃I₃O
MW: 471.8 g/mol
MDL Number: MFCD00002179
CAS Number: 609-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR