6-Chloro-2-hexanone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chlorohexan-2-one

A573562-100G A573562-25G A573562-500G A573562-5G
77117-134EA 51.5 USD
77117-134 77117-136 77117-138 77117-140
6-Chloro-2-hexanone 98%
6-Chloro-2-hexanone

Precaución: for Research Use Only

Formula: C₆H₁₁ClO
MDL Number: MFCD00191625
CAS Number: 10226-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR