1,2-Diphenyl-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A573330-1G A573330-250MG A573330-5G
77117-128EA 122.57 USD
77117-128 77117-130 77117-132
1,2-Diphenyl-1H-benzo[d]imidazole 95%
1,2-Diphenyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₉H₁₄N₂
MDL Number: MFCD00416995
CAS Number: 2622-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR