4-Nitrobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: p-Nitrobenzophenone

A572934-25G A572934-5G
77117-120EA 19.1 USD
77117-120 77117-122
4-Nitrobenzophenone 98%
4-Nitrobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉NO₃
MW: 227.22 g/mol
MDL Number: MFCD00007354
CAS Number: 1144-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR