Ethyl N-Cbz-piperidine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A571747-10G
A571747-1G
A571747-25G
A571747-5G
77117-104EA
15.45
USD
77117-104
77117-106
77117-108
77117-110
Ethyl N-Cbz-piperidine-4-carboxylate 97%
Ethyl N-Cbz-piperidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₆H₂₁NO₄ |
MDL Number:
MFCD06204513 CAS Number: 160809-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...