2'-(Trifluoromethoxy)-[1,1'-biphenyl]-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A570851-1G
A570851-250MG
77117-100EA
66.84
USD
77117-100
77117-102
2'-(Trifluoromethoxy)-[1,1'-biphenyl]-4-carboxylic acid 98%
2'-(Trifluoromethoxy)-[1,1'-biphenyl]-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₉F₃O₃ MW: 282.21 g/mol |
MDL Number:
MFCD09042195 CAS Number: 926220-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...