(5-Chloro-1H-indol-2-yl)(4-methylpiperazin-1-yl)methanone 98%

Supplier: Ambeed
Warning

Synonyms: JNJ-7777120

A568763-10MG A568763-1MG A568763-25MG A568763-50MG A568763-5MG A568763-100MG
77117-088EA 11.67 USD
77117-088 77117-090 77117-092 77117-094 77117-096 77686-451
(5-Chloro-1H-indol-2-yl)(4-methylpiperazin-1-yl)methanone 98%
(5-Chloro-1H-indol-2-yl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClN₃O
MW: 277.75 g/mol
MDL Number: MFCD04343337
CAS Number: 459168-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR