Ethyl propiolate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester
A567943-10G
A567943-1G
A567943-25G
A567943-5G
77117-080EA
14.42
USD
77117-080
77117-082
77117-084
77117-086
Ethyl propiolate 98%
Ethyl propiolate
Precaución: for Research Use Only
Formula:
C₅H₆O₂ |
MDL Number:
MFCD00009184 CAS Number: 623-47-2 UN: 3272 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...