5-Methylisoxazole 98%

Supplier: Ambeed
Danger

A567433-10G A567433-1G A567433-250MG A567433-5G
77117-072EA 111.4 USD
77117-072 77117-074 77117-076 77117-078
5-Methylisoxazole 98%
5-Methylisoxazole

Precaución: for Research Use Only

Formula: C₄H₅NO
MDL Number: MFCD00003152
CAS Number: 5765-44-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR