AK-7 98%

Supplier: Ambeed
Warning

A566314-10MG A566314-1MG A566314-25MG A566314-50MG
77117-054EA 90.63 USD
77117-054 77117-056 77117-058 77117-060
AK-7 98%
AK-7

Precaución: for Research Use Only

Formula: C₁₉H₂₁BrN₂O₃S
MDL Number: MFCD03140195
CAS Number: 420831-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR