5-Bromo-1,3-difluoro-2-isopropoxybenzene 97%

Supplier: Ambeed
Warning

A564087-100MG A564087-1G A564087-250MG
77117-028EA 14.42 USD
77117-028 77117-030 77117-032
5-Bromo-1,3-difluoro-2-isopropoxybenzene 97%
5-Bromo-1,3-difluoro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MDL Number: MFCD18917884
CAS Number: 1309933-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR