AMG-900 98%

Supplier: Ambeed
Warning

Synonyms: N-[4-[3-(2-aminopyrimidin-4-yl)pyridin-2-yl]oxyphenyl]-4-(4-methylthiophen-2-yl)phthalazin-1-amine, N-(4-((3-(2-Aminopyrimidin-4-yl)pyridin-2-yl)oxy)phenyl)-4-(4-methylthiophen-2-yl)phthalazin-1-amine, AMG 900

A563424-5MG A563424-10MG A563424-250MG A563424-50MG
77117-024EA 37.14 USD
77117-024 77594-712 77594-714 77613-758
AMG-900 98%
AMG-900

Precaución: for Research Use Only

Formula: C₂₈H₂₁N₇OS
MW: 503.58 g/mol
MDL Number: MFCD18633194
CAS Number: 945595-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR