4,6-Dibromoquinoline 95%
Supplier: Ambeed
|
Warning
|
A563107-100MG
A563107-1G
A563107-250MG
A563107-5G
77117-016EA
12.36
USD
77117-016
77117-018
77117-020
77117-022
4,6-Dibromoquinoline 95%
4,6-Dibromoquinoline
Precaución: for Research Use Only
Formula:
C₉H₅Br₂N |
MDL Number:
MFCD08436999 CAS Number: 927801-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...