2-Chloro-5-(trifluoromethyl)benzenesulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-5-(trifluoromethyl)benzene-1-sulphonyl chloride

A561592-1G A561592-25G A561592-5G
77117-010EA 11.33 USD
77117-010 77117-012 77117-014
2-Chloro-5-(trifluoromethyl)benzenesulphonyl chloride 98%
2-Chloro-5-(trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃O₂S
MW: 279.07
MDL Number: MFCD00052310
CAS Number: 54090-08-3
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR