2-Cyclohexen-1-ol 95%

Supplier: Ambeed
Danger

Synonyms: Cyclohex-2-ene-1-ol, Cyclohex-2-enol

A560939-1G A560939-250MG A560939-25G A560939-5G
77117-002EA 25.47 USD
77117-002 77117-004 77117-006 77117-008
2-Cyclohexen-1-ol 95%
2-Cyclohexen-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₀O
MW: 98.14 g/mol
MDL Number: MFCD00001570
CAS Number: 822-67-3
UN: 1987
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR