3,3-Dichloro-1-(4-iodophenyl)piperidin-2-one 95+%

Supplier: Ambeed
Warning

A559844-1G A559844-250MG A559844-5G
77116-996EA 44.56 USD
77116-996 77116-998 77117-000
3,3-Dichloro-1-(4-iodophenyl)piperidin-2-one 95+%
3,3-Dichloro-1-(4-iodophenyl)piperidin-2-one

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂INO
MW: 370.01 g/mol
MDL Number: MFCD18072443
CAS Number: 545445-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR