Sennoside A 95%

Supplier: Ambeed
Warning

Synonyms: (9R,9'R)-4,4'-Dihydroxy-10,10'-dioxo-5,5'-bis(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-9,9',10,10'-tetrahydro-[9,9'-bianthracene]-2,2'-dicarboxylic acid

A559275-1MG A559275-25MG A559275-50MG
77116-990EA 12.74 USD
77116-990 77116-992 77116-994
Sennoside A 95%
Sennoside A

Precaución: for Research Use Only

Formula: C₄₂H₃₈O₂₀
MW: 862.74 g/mol
MDL Number: MFCD00151527
CAS Number: 81-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR