H-Cys(Acm)-OH·HCl 98%

Supplier: Ambeed
Warning

Synonyms: H-Cys(Acm)-OH·HCl, S-Acetamidomethyl-L-cysteine hydrochloride, H-Cys(Acm)-OH HCl

A558542-1G A558542-250MG A558542-25G A558542-5G
77116-982EA 47.38 USD
77116-982 77116-984 77116-986 77116-988
H-Cys(Acm)-OH·HCl 98%
H-Cys(Acm)-OH·HCl

Precaución: for Research Use Only

Formula: C₆H₁₃ClN₂O₃S
MW: 228.691 g/mol
MDL Number: MFCD00077080
CAS Number: 28798-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR