4-Bromobenzyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene

A557499-10G A557499-5G A557499-25G
77116-970EA 13.8 USD
77116-970 77116-972 77594-690
4-Bromobenzyl chloride 98%
4-Bromobenzyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrCl
MW: 205.48 g/mol
MDL Number: MFCD00040867
CAS Number: 589-17-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR