N-(Triphenylmethyl)glycine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Tritylamino)acetic acid, Trt-Gly-OH, N-Tritylglycine

A555931-100G A555931-1G A555931-25G A555931-5G
77116-956EA 231.28 USD
77116-956 77116-958 77116-960 77116-962
N-(Triphenylmethyl)glycine 98%
N-(Triphenylmethyl)glycine

Precaución: for Research Use Only

Formula: C₂₁H₁₉NO₂
MW: 317.39 g/mol
MDL Number: MFCD00004276
CAS Number: 5893-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR