Ethyl-6-chloro-4-hydroxyquinoline-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 6-chloro-4-hydroxyquinoline-3-carboxylate
A555902-1G
A555902-250MG
A555902-5G
77116-950EA
45.62
USD
77116-950
77116-952
77116-954
Ethyl-6-chloro-4-hydroxyquinoline-3-carboxylate 97%
Ethyl 6-chloro-4-hydroxyquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₀ClNO₃ MW: 251.6657 g/mol |
MDL Number:
MFCD00173342 CAS Number: 70271-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...