2-Chloro-5-fluoropyrimidin-4-one 95%

Supplier: Ambeed
Warning

A554046-1G A554046-250MG A554046-5G
77116-938EA 11.34 USD
77116-938 77116-940 77686-422
2-Chloro-5-fluoropyrimidin-4-one 95%
2-Chloro-5-fluoropyrimidin-4-one

Precaución: for Research Use Only

Formula: C₄H₂ClFN₂O
MDL Number: MFCD01346689
CAS Number: 155-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR