4-Amino-3-nitrophenylboronic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-nitrophenylboronicacid

A554019-100MG A554019-250MG
77116-934EA 42.23 USD
77116-934 77116-936
4-Amino-3-nitrophenylboronic acid 98+%
4-Amino-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BN₂O₄
MDL Number: MFCD07437851
CAS Number: 89466-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR