Pentadecan-15-olide 98%

Supplier: Ambeed
Warning

Synonyms: Cyclopentadecanolide, Oxacyclohexadecan-2-one, Pentadecanolide, ω-Pentadecalactone, omega-Pentadecalactone, Oxa-2-cyclohexadecanone

A553352-100G A553352-25G A553352-500G A553352-5G
77116-926EA 54.11 USD
77116-926 77116-928 77116-930 77116-932
Pentadecan-15-olide 98%
Pentadecan-15-olide

Precaución: for Research Use Only

Formula: C₁₅H₂₈O₂
MW: 240.39 g/mol
MDL Number: MFCD00039667
CAS Number: 106-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR