3-Bromo-5-phenylsalicylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
NSC 109116, 5-PBSA, 5-Bromo-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 5-Bromo-4-hydroxy[1,1'-biphenyl]-3-carboxylic acid, AKR1C1-IN-1
A550789-10MG
A550789-1MG
A550789-50MG
77116-898EA
46.68
USD
77116-898
77116-900
77116-902
3-Bromo-5-phenylsalicylic acid 98%
3-Bromo-5-phenylsalicylic acid
Precaución: for Research Use Only
Formula:
C₁₃H₉BrO₃ MW: 293.113 g/mol |
MDL Number:
MFCD18378258 CAS Number: 4906-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...