(2-Methoxyphenyl)hydrazine hydrochloride 96%
Supplier: Ambeed
|
Warning
|
A550644-1G
A550644-25G
77116-894EA
7.21
USD
77116-894
77116-896
(2-Methoxyphenyl)hydrazine hydrochloride 96%
(2-Methoxyphenyl)hydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₁ClN₂O |
MDL Number:
MFCD00035456 CAS Number: 6971-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...