3-Bromo-2-ethoxy-5-methylbenzaldehyde 95+%

Supplier: Ambeed
Warning

A550498-1G A550498-250MG A550498-5G
77116-884EA 170.98 USD
77116-884 77116-886 77116-888
3-Bromo-2-ethoxy-5-methylbenzaldehyde 95+%
3-Bromo-2-ethoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD19105357
CAS Number: 708272-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR