Methyl (4-fluorobenzoyl)acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(4-Fluorophenyl)-3-oxopropionic Acid Methyl Ester, Methyl 4-fluorobenzoylacetate, (4-Fluorobenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Fluorophenyl)-3-oxopropionate
A547803-100G
A547803-10G
A547803-1G
A547803-25G
A547803-5G
77116-870EA
177.16
USD
77116-870
77116-872
77116-874
77116-876
77116-878
Methyl (4-fluorobenzoyl)acetate 98%
Methyl (4-fluorobenzoyl)acetate
Precaución: for Research Use Only
Formula:
C₁₀H₉FO₃ |
MDL Number:
MFCD00000355 CAS Number: 63131-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...