tert-Butyl-4-methoxy-2-methylphenylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-methoxy-2-methylphenyl)carbamate

A546043-100MG A546043-1G A546043-250MG
77116-856EA 36.08 USD
77116-856 77116-858 77116-860
tert-Butyl-4-methoxy-2-methylphenylcarbamate 95%
tert-Butyl-4-methoxy-2-methylphenylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
MW: 237.3 g/mol
MDL Number: MFCD11045029
CAS Number: 129822-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR