2,7-Naphthalenedicarboxylic acid 95%

Supplier: Ambeed
Warning

A540845-100MG A540845-1G A540845-250MG A540845-5G
77116-820EA 26.78 USD
77116-820 77116-822 77116-824 77617-302
2,7-Naphthalenedicarboxylic acid 95%
2,7-Naphthalenedicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₈O₄
MDL Number: MFCD00060862
CAS Number: 2089-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR