4-Chloro-6-(1H-imidazol-1-yl)pyrimidin-5-amine 95%

Supplier: Ambeed
Danger

A540304-100MG A540304-1G A540304-250MG
77116-814EA 20.6 USD
77116-814 77116-816 77116-818
4-Chloro-6-(1H-imidazol-1-yl)pyrimidin-5-amine 95%
4-Chloro-6-(1H-imidazol-1-yl)pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₇H₆ClN₅
MDL Number: MFCD08367609
CAS Number: 882770-78-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR