4-Chloro-3-nitrobenzoyl chloride 95%

Supplier: Ambeed
Danger

A538677-1G A538677-25G A538677-5G
77116-802EA 9.55 USD
77116-802 77116-804 77116-806
4-Chloro-3-nitrobenzoyl chloride 95%
4-Chloro-3-nitrobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₃
MW: 220.01 g/mol
MDL Number: MFCD00035743
CAS Number: 38818-50-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR