Methyl-3-furoate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl furan-3-carboxylate

A537170-1G A537170-25G A537170-5G
77116-796EA 21.22 USD
77116-796 77116-798 77116-800
Methyl-3-furoate 98%
Methyl-3-furoate

Precaución: for Research Use Only

Formula: C₆H₆O₃
MW: 126.11 g/mol
MDL Number: MFCD06203671
CAS Number: 13129-23-2
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR