WP-1130 98+%

Supplier: Ambeed
Warning

A535148-10MG A535148-1MG A535148-50MG A535148-5MG
77116-766EA 90.64 USD
77116-766 77116-768 77116-770 77116-772
WP-1130 98+%
WP-1130

Precaución: for Research Use Only

Formula: C₁₉H₁₈BrN₃O
MDL Number: MFCD18839228
CAS Number: 856243-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR