Zofenopril Calcium 98%

Supplier: Ambeed
Danger

A534063-100MG A534063-10MG A534063-1MG A534063-50MG A534063-5MG
77116-750EA 25.75 USD
77116-750 77116-752 77116-754 77116-756 77116-758
Zofenopril Calcium 98%
Zofenopril Calcium

Precaución: for Research Use Only

Formula: C₄₄H₄₄CaN₂O₈S₄
MDL Number: MFCD08704648
CAS Number: 81938-43-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR