(3,4-Dimethoxybenzyl)methylamine 95%

Supplier: Ambeed
Warning

A531810-1G A531810-250MG A531810-5G
77116-738EA 36.05 USD
77116-738 77116-740 77116-742
(3,4-Dimethoxybenzyl)methylamine 95%
(3,4-Dimethoxybenzyl)methylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD00837593
CAS Number: 63-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR